5-{[4-Chloro-3-(trifluoromethyl)-1H-pyrazol-1-yl]methyl}furan-2-carboxylic acid

Lieferant: Apollo Scientific

PC410409-1G
APOSPC410409-1GEA 232 EUR
APOSPC410409-1G
5-{[4-Chloro-3-(trifluoromethyl)-1H-pyrazol-1-yl]methyl}furan-2-carboxylic acid
5-{[4-Chloro-3-(trifluoromethyl)-1H-pyrazol-1-yl]methyl}furan-2-carboxylic acid
Formel: C₁₀H₆ClF₃N₂O₃
Molecular Weight: 294,614 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04970080
CAS-Nummer: 1006348-61-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR