4-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]aniline

Lieferant: Apollo Scientific

PC410410-1G
APOSPC410410-1GEA 647 EUR
APOSPC410410-1G
4-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]aniline
4-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]aniline
Formel: C₁₁H₁₀F₃N₃
Molecular Weight: 241.212 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08558191
CAS-Nummer: 1006348-64-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR