3-[1',5'-Dimethyl-5-(trifluoromethyl)-1'H,2H-3,4'-bipyrazol-2-yl]propanenitrile

Lieferant: Apollo Scientific

PC410411-1G
APOSPC410411-1GEA 386 EUR
APOSPC410411-1G
3-[1',5'-Dimethyl-5-(trifluoromethyl)-1'H,2H-3,4'-bipyrazol-2-yl]propanenitrile
3-[1',5'-Dimethyl-5-(trifluoromethyl)-1'H,2H-3,4'-bipyrazol-2-yl]propanenitrile
Formel: C₁₂H₁₂F₃N₅
Molecular Weight: 283,252 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08559023
CAS-Nummer: 1006348-67-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR