4-Methyl-5-[3-(trifluoromethyl)-1H-pyrazol-1-yl]-1,3-thiazol-2-amine
Lieferant: Apollo Scientific
PC410412-1G
APOSPC410412-1GEA
516
EUR
APOSPC410412-1G
4-Methyl-5-[3-(trifluoromethyl)-1H-pyrazol-1-yl]-1,3-thiazol-2-amine
4-Methyl-5-[3-(trifluoromethyl)-1H-pyrazol-1-yl]-1,3-thiazol-2-amine
Formel:
C₈H₇F₃N₄S Molecular Weight: 248,228 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08558209 CAS-Nummer: 1006348-68-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...