[3-(Trifluoromethyl)-1H-pyrazol-1-yl]acetonitrile

Lieferant: Apollo Scientific

PC410413-1G
APOSPC410413-1GEA 516 EUR
APOSPC410413-1G
[3-(Trifluoromethyl)-1H-pyrazol-1-yl]acetonitrile
[3-(Trifluoromethyl)-1H-pyrazol-1-yl]acetonitrile
Formel: C₆H₄F₃N₃
Molecular Weight: 175,111 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04969709
CAS-Nummer: 1006348-71-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR