2-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanoic acid

Lieferant: Apollo Scientific

PC410414-1G
APOSPC410414-1GEA 386 EUR
APOSPC410414-1G
2-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanoic acid
2-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanoic acid
Formel: C₁₁H₁₃F₃N₂O₂
Molecular Weight: 262,228 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04967804
CAS-Nummer: 1006348-77-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR