2-Methyl-3-[3-methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]propan-1-amine

Lieferant: Apollo Scientific

PC410415-1G
APOSPC410415-1GEA 183 EUR
APOSPC410415-1G
2-Methyl-3-[3-methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]propan-1-amine
2-Methyl-3-[3-methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]propan-1-amine
Formel: C₉H₁₄F₃N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD08559043
CAS-Nummer: 1006348-75-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR