1-[4-(1,5-Dimethyl-1H-pyrazol-4-yl)-6-(trifluoromethyl)pyrimidin-2-yl]piperidine-3-carboxylic acid

Lieferant: Apollo Scientific

PC410416-1G
APOSPC410416-1GEA 647 EUR
APOSPC410416-1G
1-[4-(1,5-Dimethyl-1H-pyrazol-4-yl)-6-(trifluoromethyl)pyrimidin-2-yl]piperidine-3-carboxylic acid
1-[4-(1,5-Dimethyl-1H-pyrazol-4-yl)-6-(trifluoromethyl)pyrimidin-2-yl]piperidine-3-carboxylic acid
Formel: C₁₆H₁₈F₃N₅O₂
Molecular Weight: 369,342 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05861950
CAS-Nummer: 1006348-78-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR