3-[4-Bromo-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid

Lieferant: Apollo Scientific

PC410417-1G
APOSPC410417-1GEA 647 EUR
APOSPC410417-1G
3-[4-Bromo-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
3-[4-Bromo-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
Formel: C₇H₆BrF₃N₂O₂
Molecular Weight: 287,034 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04969244
CAS-Nummer: 1006348-79-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR