5-Methyl-1-{[5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]methyl}-1H-pyrazol-3-amine
Lieferant: Apollo Scientific
PC410419-1G
APOSPC410419-1GEA
0
EUR
APOSPC410419-1G
5-Methyl-1-{[5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]methyl}-1H-pyrazol-3-amine
5-Methyl-1-{[5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]methyl}-1H-pyrazol-3-amine
Formel:
C₁₀H₁₂F₃N₅ Lagertemperatur: Raumtemperatur |
MDL:
MFCD04967743 CAS-Nummer: 1006349-20-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...