(1E)-N'-Hydroxy-2-methyl-3-[5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanimidamide

Lieferant: Apollo Scientific

PC410420-1G
APOSPC410420-1GEA 386 EUR
APOSPC410420-1G
(1E)-N'-Hydroxy-2-methyl-3-[5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanimidamide
(1E)-N'-Hydroxy-2-methyl-3-[5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanimidamide
Formel: C₉H₁₃F₃N₄O
Molecular Weight: 250,221 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08556213
CAS-Nummer: 1006352-70-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR