1-(1-Ethyl-3-methyl-1H-pyrazol-4-yl)-N-(4-fluorobenzyl)methanamine
Lieferant: Apollo Scientific
PC410421-1G
APOSPC410421-1GEA
647
EUR
APOSPC410421-1G
1-(1-Ethyl-3-methyl-1H-pyrazol-4-yl)-N-(4-fluorobenzyl)methanamine
1-(1-Ethyl-3-methyl-1H-pyrazol-4-yl)-N-(4-fluorobenzyl)methanamine
Formel:
C₁₄H₁₈FN₃ Molecular Weight: 247,311 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04968223 CAS-Nummer: 1006352-79-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...