(1Z)-N'-Hydroxy-3-[5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanimidamide

Lieferant: Apollo Scientific

PC410424-1G
APOSPC410424-1GEA 386 EUR
APOSPC410424-1G
(1Z)-N'-Hydroxy-3-[5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanimidamide
(1Z)-N'-Hydroxy-3-[5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanimidamide
Formel: C₈H₁₁F₃N₄O
Molecular Weight: 236,194 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15976230
CAS-Nummer: 1006353-07-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR