3-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]butan-1-amine

Lieferant: Apollo Scientific

PC410426-1G
APOSPC410426-1GEA 516 EUR
APOSPC410426-1G
3-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]butan-1-amine
3-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]butan-1-amine
Formel: C₁₁H₁₆F₃N₃
Molecular Weight: 247,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06805095
CAS-Nummer: 1006353-19-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR