(1Z)-N'-Hydroxy-2-[5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]ethanimidamide

Lieferant: Apollo Scientific

PC410427-1G
APOSPC410427-1GEA 516 EUR
APOSPC410427-1G
(1Z)-N'-Hydroxy-2-[5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]ethanimidamide
(1Z)-N'-Hydroxy-2-[5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]ethanimidamide
Formel: C₇H₉F₃N₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD08557063
CAS-Nummer: 1006354-87-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR