1-(Chloromethyl)-5-methyl-3-(trifluoromethyl)-1H-pyrazole

Lieferant: Apollo Scientific

PC410428-1G
APOSPC410428-1GEA 647 EUR
APOSPC410428-1G
1-(Chloromethyl)-5-methyl-3-(trifluoromethyl)-1H-pyrazole
1-(Chloromethyl)-5-methyl-3-(trifluoromethyl)-1H-pyrazole
Formel: C₆H₆ClF₃N₂
Molecular Weight: 198,573 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04968631
CAS-Nummer: 1006356-44-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR