3-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]-2-methylpropanenitrile

Lieferant: Apollo Scientific

PC410429-1G
APOSPC410429-1GEA 386 EUR
APOSPC410429-1G
3-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]-2-methylpropanenitrile
3-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]-2-methylpropanenitrile
Formel: C₁₁H₁₂F₃N₃
Molecular Weight: 243,228 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06739345
CAS-Nummer: 1006356-58-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR