4-Nitro-1-(2,2,2-trifluoroethyl)-1H-pyrazole-3-carboxylic acid
Lieferant: Apollo Scientific
PC410431-1G
APOSPC410431-1GEA
1080
EUR
APOSPC410431-1G
4-Nitro-1-(2,2,2-trifluoroethyl)-1H-pyrazole-3-carboxylic acid
4-Nitro-1-(2,2,2-trifluoroethyl)-1H-pyrazole-3-carboxylic acid
Formel:
C₆H₄F₃N₃O₄ Molecular Weight: 239,109 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08696333 CAS-Nummer: 1006433-24-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...