5-(1,3-Dimethyl-1H-pyrazol-4-yl)-7-(trifluoromethyl)-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine

Lieferant: Apollo Scientific

PC410432-1G
APOSPC410432-1GEA 828 EUR
APOSPC410432-1G
5-(1,3-Dimethyl-1H-pyrazol-4-yl)-7-(trifluoromethyl)-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine
5-(1,3-Dimethyl-1H-pyrazol-4-yl)-7-(trifluoromethyl)-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine
Formel: C₁₂H₁₄F₃N₅
Molecular Weight: 285.268 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04968855
CAS-Nummer: 1006434-36-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR