4-Chloro-1-[(2-fluorophenoxy)methyl]-1H-pyrazol-3-amine
Lieferant: Apollo Scientific
PC410433-1G
APOSPC410433-1GEA
516
EUR
APOSPC410433-1G
4-Chloro-1-[(2-fluorophenoxy)methyl]-1H-pyrazol-3-amine
4-Chloro-1-[(2-fluorophenoxy)methyl]-1H-pyrazol-3-amine
Formel:
C₁₀H₉ClFN₃O Molecular Weight: 241,649 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04967547 CAS-Nummer: 1006435-98-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...