1-(3,5-Difluorophenyl)-N-[(1-methyl-1H-pyrazol-4-yl)methyl]methanamine
Lieferant: Apollo Scientific
PC410434-1G
APOSPC410434-1GEA
828
EUR
APOSPC410434-1G
1-(3,5-Difluorophenyl)-N-[(1-methyl-1H-pyrazol-4-yl)methyl]methanamine
1-(3,5-Difluorophenyl)-N-[(1-methyl-1H-pyrazol-4-yl)methyl]methanamine
Formel:
C₁₂H₁₃F₂N₃ Molecular Weight: 237,249 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08696781 CAS-Nummer: 1006436-32-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...