2-(3-Trifluoromethylpyrazol-1-yl)ethylamine
Lieferant: Apollo Scientific
|
Achtung
|
PC410435-1G
PC410435-5G
APOSPC410435-1GEA
453
EUR
APOSPC410435-1G
APOSPC410435-5G
2-(3-Trifluoromethylpyrazol-1-yl)ethylamine
2-(3-Trifluoromethylpyrazol-1-yl)ethylamine
Formel:
C₆H₈F₃N₃ Molecular Weight: 179.15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04970067 CAS-Nummer: 1006436-51-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...