1-(3,5-Difluorophenyl)-N-[(1,3-dimethyl-1H-pyrazol-4-yl)methyl]methanamine
Lieferant: Apollo Scientific
PC410436-1G
APOSPC410436-1GEA
828
EUR
APOSPC410436-1G
1-(3,5-Difluorophenyl)-N-[(1,3-dimethyl-1H-pyrazol-4-yl)methyl]methanamine
1-(3,5-Difluorophenyl)-N-[(1,3-dimethyl-1H-pyrazol-4-yl)methyl]methanamine
Formel:
C₁₃H₁₅F₂N₃ Molecular Weight: 251,275 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08696806 CAS-Nummer: 1006436-76-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...