{[4-(1-Methyl-1H-pyrazol-4-yl)-6-(trifluoromethyl)pyrimidin-2-yl]sulphanyl}acetic acid

Lieferant: Apollo Scientific

PC410438-1G
APOSPC410438-1GEA 647 EUR
APOSPC410438-1G
{[4-(1-Methyl-1H-pyrazol-4-yl)-6-(trifluoromethyl)pyrimidin-2-yl]sulphanyl}acetic acid
{[4-(1-Methyl-1H-pyrazol-4-yl)-6-(trifluoromethyl)pyrimidin-2-yl]sulphanyl}acetic acid
Formel: C₁₁H₉F₃N₄O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD04970366
CAS-Nummer: 1006440-64-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR