1-[2-Nitro-4-(trifluoromethyl)phenyl]-1H-pyrazole-3-carboxylic acid

Lieferant: Apollo Scientific

PC410440-1G
APOSPC410440-1GEA 647 EUR
APOSPC410440-1G
1-[2-Nitro-4-(trifluoromethyl)phenyl]-1H-pyrazole-3-carboxylic acid
1-[2-Nitro-4-(trifluoromethyl)phenyl]-1H-pyrazole-3-carboxylic acid
Formel: C₁₁H₆F₃N₃O₄
Molecular Weight: 301,178 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08696945
CAS-Nummer: 1006442-79-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR