1-(4-Fluoro-2-nitrophenyl)-1H-pyrazole-3-carboxylic acid
Lieferant: Apollo Scientific
PC410441-1G
APOSPC410441-1GEA
647
EUR
APOSPC410441-1G
1-(4-Fluoro-2-nitrophenyl)-1H-pyrazole-3-carboxylic acid
1-(4-Fluoro-2-nitrophenyl)-1H-pyrazole-3-carboxylic acid
Formel:
C₁₀H₆FN₃O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD08696948 CAS-Nummer: 1006442-84-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...