1-(2-Fluoro-4-nitrophenyl)-1H-pyrazole-3-carboxylic acid

Lieferant: Apollo Scientific

PC410442-1G
APOSPC410442-1GEA 647 EUR
APOSPC410442-1G
1-(2-Fluoro-4-nitrophenyl)-1H-pyrazole-3-carboxylic acid
1-(2-Fluoro-4-nitrophenyl)-1H-pyrazole-3-carboxylic acid
Formel: C₁₀H₆FN₃O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD08696951
CAS-Nummer: 1006442-88-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR