1-(2,2-Difluoroethyl)-3,5-dimethyl-4-nitro-1H-pyrazole

Lieferant: Apollo Scientific

PC410443-1G
APOSPC410443-1GEA 386 EUR
APOSPC410443-1G
1-(2,2-Difluoroethyl)-3,5-dimethyl-4-nitro-1H-pyrazole
1-(2,2-Difluoroethyl)-3,5-dimethyl-4-nitro-1H-pyrazole
Formel: C₇H₉F₂N₃O₂
Molecular Weight: 205,162 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08558993
CAS-Nummer: 1006443-08-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR