[6-(1-Ethyl-1H-pyrazol-4-yl)-3-methyl-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid
Lieferant: Apollo Scientific
PC410444-1G
APOSPC410444-1GEA
647
EUR
APOSPC410444-1G
[6-(1-Ethyl-1H-pyrazol-4-yl)-3-methyl-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid
[6-(1-Ethyl-1H-pyrazol-4-yl)-3-methyl-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid
Formel:
C₁₅H₁₄F₃N₅O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD08697654 CAS-Nummer: 1006444-20-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...