[3-Cyclopropyl-6-(1,5-dimethyl-1H-pyrazol-4-yl)-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid

Lieferant: Apollo Scientific

PC410445-1G
APOSPC410445-1GEA 828 EUR
APOSPC410445-1G
[3-Cyclopropyl-6-(1,5-dimethyl-1H-pyrazol-4-yl)-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid
[3-Cyclopropyl-6-(1,5-dimethyl-1H-pyrazol-4-yl)-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid
Formel: C₁₇H₁₆F₃N₅O₂
Molecular Weight: 379,336 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08697673
CAS-Nummer: 1006444-33-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR