[4-(Difluoromethyl)-3-methyl-6-(1-methyl-1H-pyrazol-4-yl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid

Lieferant: Apollo Scientific

PC410446-1G
APOSPC410446-1GEA 828 EUR
APOSPC410446-1G
[4-(Difluoromethyl)-3-methyl-6-(1-methyl-1H-pyrazol-4-yl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid
[4-(Difluoromethyl)-3-methyl-6-(1-methyl-1H-pyrazol-4-yl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid
Formel: C₁₄H₁₃F₂N₅O₂
Molecular Weight: 321,282 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08697724
CAS-Nummer: 1006444-93-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR