[4-(Difluoromethyl)-6-(1,3-dimethyl-1H-pyrazol-4-yl)-3-methyl-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid
Lieferant: Apollo Scientific
PC410447-1G
APOSPC410447-1GEA
828
EUR
APOSPC410447-1G
[4-(Difluoromethyl)-6-(1,3-dimethyl-1H-pyrazol-4-yl)-3-methyl-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid
[4-(Difluoromethyl)-6-(1,3-dimethyl-1H-pyrazol-4-yl)-3-methyl-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid
Formel:
C₁₅H₁₅F₂N₅O₂ Molecular Weight: 335.309 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08697729 CAS-Nummer: 1006444-98-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...