[3-Cyclopropyl-4-(difluoromethyl)-6-(1,3-dimethyl-1H-pyrazol-4-yl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid

Lieferant: Apollo Scientific

PC410449-1G
APOSPC410449-1GEA 980 EUR
APOSPC410449-1G
[3-Cyclopropyl-4-(difluoromethyl)-6-(1,3-dimethyl-1H-pyrazol-4-yl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid
[3-Cyclopropyl-4-(difluoromethyl)-6-(1,3-dimethyl-1H-pyrazol-4-yl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid
Formel: C₁₇H₁₇F₂N₅O₂
Molecular Weight: 361.346 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08697749
CAS-Nummer: 1006445-13-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR