[4-Chloro-3-(trifluoromethyl)-1H-pyrazol-1-yl]acetic acid

Lieferant: Apollo Scientific

PC410450-1G
APOSPC410450-1GEA 0 EUR
APOSPC410450-1G
[4-Chloro-3-(trifluoromethyl)-1H-pyrazol-1-yl]acetic acid
[4-Chloro-3-(trifluoromethyl)-1H-pyrazol-1-yl]acetic acid
Formel: C₆H₄ClF₃N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04969232
CAS-Nummer: 1006446-52-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR