[4-Chloro-5-cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]acetic acid

Lieferant: Apollo Scientific

PC410452-1G
APOSPC410452-1GEA 647 EUR
APOSPC410452-1G
[4-Chloro-5-cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]acetic acid
[4-Chloro-5-cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]acetic acid
Formel: C₉H₈ClF₃N₂O₂
Molecular Weight: 268.62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03419740
CAS-Nummer: 1006447-79-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR