1-[4-Cyclopropyl-6-(trifluoromethyl)pyrimidin-2-yl]-1H-pyrazole-3-carboxylic acid
Lieferant: Apollo Scientific
PC410453-1G
APOSPC410453-1GEA
647
EUR
APOSPC410453-1G
1-[4-Cyclopropyl-6-(trifluoromethyl)pyrimidin-2-yl]-1H-pyrazole-3-carboxylic acid
1-[4-Cyclopropyl-6-(trifluoromethyl)pyrimidin-2-yl]-1H-pyrazole-3-carboxylic acid
Formel:
C₁₂H₉F₃N₄O₂ Molecular Weight: 298.221 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05861884 CAS-Nummer: 1006448-08-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...