N-[(1-Methyl-1H-pyrazol-3-yl)methyl]-2-(trifluoromethyl)aniline

Lieferant: Apollo Scientific

PC410454-1G
APOSPC410454-1GEA 980 EUR
APOSPC410454-1G
N-[(1-Methyl-1H-pyrazol-3-yl)methyl]-2-(trifluoromethyl)aniline
N-[(1-Methyl-1H-pyrazol-3-yl)methyl]-2-(trifluoromethyl)aniline
Formel: C₁₂H₁₂F₃N₃
Molecular Weight: 255.239 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04968467
CAS-Nummer: 1006449-98-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR