3-[4-Chloro-5-cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propan-1-amine

Lieferant: Apollo Scientific

PC410455-1G
APOSPC410455-1GEA 1080 EUR
APOSPC410455-1G
3-[4-Chloro-5-cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propan-1-amine
3-[4-Chloro-5-cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propan-1-amine
Formel: C₁₀H₁₃ClF₃N₃
Molecular Weight: 267,679 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03422564
CAS-Nummer: 1006454-49-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR