3-[4-Nitro-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid

Lieferant: Apollo Scientific
Achtung

PC410459-1G
APOSPC410459-1GEA 828 EUR
APOSPC410459-1G
3-[4-Nitro-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
3-[4-Nitro-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
Formel: C₇H₆F₃N₃O₄
Molecular Weight: 253,135 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04970061
CAS-Nummer: 1006459-41-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR