2-[4-Nitro-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanoic acid

Lieferant: Apollo Scientific

PC410460-1G
APOSPC410460-1GEA 0 EUR
APOSPC410460-1G
2-[4-Nitro-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanoic acid
2-[4-Nitro-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanoic acid
Formel: C₈H₈F₃N₃O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD04970066
CAS-Nummer: 1006459-45-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR