3-[3-Methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]propanenitrile

Lieferant: Apollo Scientific

PC410461-1G
APOSPC410461-1GEA 386 EUR
APOSPC410461-1G
3-[3-Methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]propanenitrile
3-[3-Methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]propanenitrile
Formel: C₈H₈F₃N₃
Molecular Weight: 203,164 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08558151
CAS-Nummer: 1006461-11-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR