3-[1'-Methyl-5-(trifluoromethyl)-1'H,2H-3,4'-bipyrazol-2-yl]propanenitrile

Lieferant: Apollo Scientific

PC410462-1G
APOSPC410462-1GEA 516 EUR
APOSPC410462-1G
3-[1'-Methyl-5-(trifluoromethyl)-1'H,2H-3,4'-bipyrazol-2-yl]propanenitrile
3-[1'-Methyl-5-(trifluoromethyl)-1'H,2H-3,4'-bipyrazol-2-yl]propanenitrile
Formel: C₁₁H₁₀F₃N₅
Molecular Weight: 269,226 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08696393
CAS-Nummer: 1006461-64-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR