1-(2,2-Difluoroethyl)-1H-pyrazol-3-amine

Lieferant: Apollo Scientific

PC410464-1G
APOSPC410464-1GEA 154 EUR
APOSPC410464-1G
1-(2,2-Difluoroethyl)-1H-pyrazol-3-amine
1-(2,2-Difluoroethyl)-1H-pyrazol-3-amine
Formel: C₅H₇F₂N₃
Molecular Weight: 147,126 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08696542
CAS-Nummer: 1006462-38-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR