1-(4-Fluorophenyl)-N-[(1-methyl-1H-pyrazol-4-yl)methyl]methanamine
Lieferant: Apollo Scientific
PC410465-1G
APOSPC410465-1GEA
828
EUR
APOSPC410465-1G
1-(4-Fluorophenyl)-N-[(1-methyl-1H-pyrazol-4-yl)methyl]methanamine
1-(4-Fluorophenyl)-N-[(1-methyl-1H-pyrazol-4-yl)methyl]methanamine
Formel:
C₁₂H₁₄FN₃ Molecular Weight: 219,258 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04968191 CAS-Nummer: 1006464-87-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...