1-(1,3-Dimethyl-1H-pyrazol-4-yl)-N-(4-fluorobenzyl)methanamine

Lieferant: Apollo Scientific

PC410466-1G
APOSPC410466-1GEA 828 EUR
APOSPC410466-1G
1-(1,3-Dimethyl-1H-pyrazol-4-yl)-N-(4-fluorobenzyl)methanamine
1-(1,3-Dimethyl-1H-pyrazol-4-yl)-N-(4-fluorobenzyl)methanamine
Formel: C₁₃H₁₆FN₃
Molecular Weight: 233,285 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04968199
CAS-Nummer: 1006464-99-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR