1-(1,5-Dimethyl-1H-pyrazol-4-yl)-N-(4-fluorobenzyl)methanamine
Lieferant: Apollo Scientific
PC410467-1G
APOSPC410467-1GEA
828
EUR
APOSPC410467-1G
1-(1,5-Dimethyl-1H-pyrazol-4-yl)-N-(4-fluorobenzyl)methanamine
1-(1,5-Dimethyl-1H-pyrazol-4-yl)-N-(4-fluorobenzyl)methanamine
Formel:
C₁₃H₁₆FN₃ Molecular Weight: 233.285 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04968207 CAS-Nummer: 1006465-11-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...