3-Chloro-4-[3-(trifluoromethyl)-1H-pyrazol-1-yl]aniline

Lieferant: Apollo Scientific

PC410468-1G
APOSPC410468-1GEA 828 EUR
APOSPC410468-1G
3-Chloro-4-[3-(trifluoromethyl)-1H-pyrazol-1-yl]aniline
3-Chloro-4-[3-(trifluoromethyl)-1H-pyrazol-1-yl]aniline
Formel: C₁₀H₇ClF₃N₃
Molecular Weight: 261,631 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08699366
CAS-Nummer: 1006467-61-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR