5-Fluoro-2-(4-methyl-1H-pyrazol-1-yl)aniline
Lieferant: Apollo Scientific
PC410469-1G
APOSPC410469-1GEA
516
EUR
APOSPC410469-1G
5-Fluoro-2-(4-methyl-1H-pyrazol-1-yl)aniline
5-Fluoro-2-(4-methyl-1H-pyrazol-1-yl)aniline
Formel:
C₁₀H₁₀FN₃ Molecular Weight: 191,205 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08699511 CAS-Nummer: 1006468-17-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...