2-(4-Chloro-1H-pyrazol-1-yl)-5-fluoroaniline
Lieferant: Apollo Scientific
PC410470-1G
APOSPC410470-1GEA
516
EUR
APOSPC410470-1G
2-(4-Chloro-1H-pyrazol-1-yl)-5-fluoroaniline
2-(4-Chloro-1H-pyrazol-1-yl)-5-fluoroaniline
Formel:
C₉H₇ClFN₃ Molecular Weight: 211,623 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08699513 CAS-Nummer: 1006468-20-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...