5-Fluoro-2-[(1-methyl-1H-pyrazol-4-yl)methoxy]aniline

Lieferant: Apollo Scientific

PC410471-1G
APOSPC410471-1GEA 980 EUR
APOSPC410471-1G
5-Fluoro-2-[(1-methyl-1H-pyrazol-4-yl)methoxy]aniline
5-Fluoro-2-[(1-methyl-1H-pyrazol-4-yl)methoxy]aniline
Formel: C₁₁H₁₂FN₃O
Molecular Weight: 221,231 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08699522
CAS-Nummer: 1006468-29-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR